Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102762.1 | NP_001096232.1 | 6 - 7 | 632 | 110 | |
GenBank mRNA | BC135340.1 | - | 6 - 7 | 632 | 110 | |
BC155857.1 | - | 6 - 7 | 610 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |