Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102778.1 | NP_001096248.1 | 3 - 4 | 792 | 57 | |
GenBank mRNA | BC135402.1 | - | 3 - 4 | 792 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |