Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102785.1 | NP_001096255.1 | 5 - 6 | 1259 | 100 | |
XM_012969954.1 | XP_012825408.1 | 5 - 6 | 1242 | 100 | ||
GenBank mRNA | BC135429.1 | - | 5 - 6 | 1259 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |