Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102831.1 | NP_001096301.1 | 3 - 4 | 1690 | 85 | |
GenBank mRNA | BC135631.1 | - | 3 - 4 | 1690 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |