Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102833.1 | NP_001096303.1 | 13 - 14 | 1331 | 95 | |
GenBank mRNA | BC135644.1 | - | 13 - 14 | 1331 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |