Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102850.1 | NP_001096320.1 | 3 - 4 | 758 | 99 | |
GenBank mRNA | BC127552.1 | - | 3 - 4 | 758 | 99 | |
BC135712.1 | - | 3 - 4 | 742 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |