Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldlrad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102860.1 | NP_001096330.1 | 3 - 4 | 803 | 88 | |
GenBank mRNA | BC064324.1 | - | 3 - 4 | 785 | 88 | |
BC135753.1 | - | 3 - 4 | 803 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |