Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102867.1 | NP_001096337.1 | 1 - 2 | 278 | 94 | |
GenBank mRNA | BC135774.1 | - | 1 - 2 | 278 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |