Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018089411.1 | XP_017944900.1 | 6 - 7 | 2736 | 71 | |
GenBank mRNA | BC135929.1 | - | 6 - 7 | 1374 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |