Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trmt44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012955724.2 | XP_012811178.1 | 4 - 5 | 892 | 99 | |
XM_012955725.2 | XP_012811179.1 | 4 - 5 | 922 | 99 | ||
GenBank mRNA | BC135974.1 | - | 4 - 5 | 840 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |