Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102926.1 | NP_001096396.1 | 1 | 1752 | 92 | |
GenBank mRNA | BC136011.1 | - | 1 | 1752 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |