Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adcyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102955.1 | NP_001096425.1 | 3 - 4 | 612 | 76 | |
XM_012964196.2 | XP_012819650.1 | 3 - 4 | 1071 | 76 | ||
GenBank mRNA | BC136094.1 | - | 3 - 4 | 612 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |