Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cryba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103017.1 | NP_001096487.1 | 3 - 4 | 193 | 76 | |
XM_012967013.2 | XP_012822467.1 | 2 - 3 | 146 | 76 | ||
GenBank mRNA | BC135346.1 | - | 3 - 4 | 193 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |