Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103048.1 | NP_001096518.1 | 2 - 3 | 1074 | 66 | |
GenBank mRNA | BC135591.1 | - | 2 - 3 | 1074 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |