Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptrhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103060.1 | NP_001096530.1 | 1 - 2 | 199 | 84 | |
GenBank mRNA | BC135811.1 | - | 1 - 2 | 199 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |