Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103063.1 | NP_001096533.1 | 5 - 6 | 315 | 75 | |
GenBank mRNA | BC135875.1 | - | 5 - 6 | 315 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |