Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c12orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016016.3 | NP_001016016.1 | 2 - 3 | 313 | 101 | |
GenBank mRNA | BC166293.1 | - | 2 - 3 | 313 | 101 | |
BC170669.1 | - | 2 - 3 | 637 | 101 | ||
BC170671.1 | - | 2 - 3 | 637 | 101 | ||
CR848210.2 | - | 2 - 3 | 658 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |