Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016278.3 | NP_001016278.2 | 1012 | 102 | ||
GenBank mRNA | BC158423.1 | - | 1012 | 102 | ||
CR761613.2 | - | 1006 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis |