Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017134.3 | NP_001017134.1 | 13 - 14 | 1325 | 88 | |
GenBank mRNA | CR761047.3 | - | 13 - 14 | 1325 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |