Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017134.3 | NP_001017134.1 | 3 - 4 | 495 | 103 | |
GenBank mRNA | CR761047.3 | - | 3 - 4 | 495 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |