Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103077.1 | NP_001096547.1 | 6 - 7 | 861 | 167 | |
XM_018094682.1 | XP_017950171.1 | 5 - 6 | 795 | 167 | ||
XM_018094695.1 | XP_017950184.1 | 5 - 6 | 798 | 164 | ||
XM_018094742.1 | XP_017950231.1 | 5 - 6 | 793 | 167 | ||
GenBank mRNA | BC136181.1 | - | 6 - 7 | 861 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |