Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103081.1 | NP_001096551.1 | 3 - 4 | 666 | 99 | |
GenBank mRNA | DQ658157.1 | - | 3 - 4 | 666 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |