Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103172.1 | NP_001096642.1 | 4 - 5 | 425 | 65 | |
XM_018096124.1 | XP_017951613.1 | 3 - 4 | 383 | 65 | ||
GenBank mRNA | CT030574.1 | - | 4 - 5 | 425 | 65 |
Gene Symbol: LOC100490060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002931800.4 | XP_002931846.1 | 4 - 5 | 458 | 65 | |
XM_018096602.1 | XP_017952091.1 | 3 - 4 | 385 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |