Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105514.1 | NP_001098984.1 | 1 - 2 | 222 | 58 | |
GenBank mRNA | CT030066.2 | - | 1 - 2 | 305 | 58 | |
CU025129.1 | - | 1 - 2 | 222 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |