Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110052.1 | NP_001103522.1 | 3 - 4 | 582 | 76 | |
XM_012968030.1 | XP_012823484.1 | 3 - 4 | 603 | 76 | ||
GenBank mRNA | BC153718.1 | - | 3 - 4 | 582 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |