Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112907.1 | NP_001106378.1 | 4 - 5 | 1031 | 70 | |
GenBank mRNA | BC155509.1 | - | 4 - 5 | 1031 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |