Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsnare1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112923.1 | NP_001106394.1 | 4 - 5 | 364 | 105 | |
GenBank mRNA | BC152038.1 | - | 4 - 5 | 364 | 105 | |
CT025175.1 | - | 5 - 6 | 404 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |