Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112955.1 | NP_001106426.1 | 9 - 10 | 2258 | 103 | |
GenBank mRNA | BC153370.1 | - | 9 - 10 | 2258 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |