Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112959.1 | NP_001106430.1 | 1 - 2 | 166 | 101 | |
GenBank mRNA | BC153710.1 | - | 1 - 2 | 166 | 101 | |
BC158169.1 | - | 1 - 2 | 168 | 101 | ||
CR761930.2 | - | 1 - 2 | 165 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |