Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112968.2 | NP_001106439.2 | 2 - 3 | 590 | 67 | |
GenBank mRNA | BC153728.1 | - | 2 - 3 | 583 | 67 | |
CX365814.2 | - | 2 - 3 | 591 | 67 | ||
DN036490.1 | - | 1 - 2 | 106 | 67 | ||
EL834187.1 | - | 2 - 3 | 590 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |