Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112988.1 | NP_001106459.1 | 5 - 6 | 737 | 67 | |
GenBank mRNA | BC154071.1 | - | 5 - 6 | 737 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |