Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113074.1 | NP_001106545.1 | 7 - 8 | 877 | 57 | |
GenBank mRNA | BC154922.1 | - | 7 - 8 | 877 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |