Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: birc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113122.1 | NP_001106593.2 | 7 - 8 | 1255 | 76 | |
GenBank mRNA | BC155423.1 | - | 7 - 8 | 1255 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |