Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ocel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113149.1 | NP_001106620.1 | 3 - 4 | 905 | 67 | |
XM_012971465.2 | XP_012826919.1 | 3 - 4 | 1571 | 67 | ||
XM_012971475.2 | XP_012826929.1 | 3 - 4 | 1565 | 67 | ||
XM_012971512.2 | XP_012826966.1 | 3 - 4 | 986 | 67 | ||
GenBank mRNA | BC155467.1 | - | 3 - 4 | 905 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |