Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113667.2 | NP_001107139.2 | 1 | 670 | 76 | |
GenBank mRNA | BC157248.1 | - | 1 | 651 | 76 | |
CU025011.1 | - | 1 | 423 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |