Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113672.1 | NP_001107144.1 | 3 - 4 | 359 | 69 | |
XM_012960554.2 | XP_012816008.1 | 3 - 4 | 717 | 69 | ||
GenBank mRNA | BC155999.1 | - | 3 - 4 | 359 | 69 | |
BC170810.1 | - | 3 - 4 | 352 | 69 | ||
BC170812.1 | - | 3 - 4 | 348 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |