Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113684.1 | NP_001107156.1 | 10 - 11 | 1181 | 59 | |
XM_012967939.2 | XP_012823393.1 | 11 - 12 | 1223 | 59 | ||
GenBank mRNA | BC155689.1 | - | 10 - 11 | 1181 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |