Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113685.1 | NP_001107157.1 | 6 - 7 | 655 | 100 | |
GenBank mRNA | BC157556.1 | - | 6 - 7 | 655 | 100 | |
BC170945.1 | - | 6 - 7 | 655 | 100 | ||
BC170947.1 | - | 6 - 7 | 655 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |