Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113688.1 | NP_001107160.1 | 12 - 13 | 2014 | 69 | |
GenBank mRNA | BC155677.1 | - | 12 - 13 | 2014 | 69 | |
BC170858.1 | - | 12 - 13 | 1889 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |