Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113830.1 | NP_001107302.1 | 1049 | 105 | ||
GenBank mRNA | BC155697.1 | - | 1049 | 105 | ||
CR942773.1 | - | 783 | 105 | |||
CU464102.1 | - | 1045 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis |