Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113840.1 | NP_001107312.1 | 8 - 9 | 1549 | 102 | |
GenBank mRNA | BC155709.1 | - | 8 - 9 | 1549 | 102 | |
BC171100.1 | - | 9 - 10 | 1402 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |