Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113881.1 | NP_001107353.1 | 5 - 6 | 445 | 97 | |
XM_012960561.2 | XP_012816015.1 | 5 - 6 | 509 | 97 | ||
XM_012960562.2 | XP_012816016.1 | 5 - 6 | 641 | 97 | ||
GenBank mRNA | BC157195.1 | - | 5 - 6 | 445 | 97 | |
BC171197.1 | - | 5 - 6 | 422 | 97 | ||
BC171199.1 | - | 5 - 6 | 422 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |