Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113882.1 | NP_001107354.1 | 3 - 4 | 377 | 83 | |
GenBank mRNA | BC157201.1 | - | 3 - 4 | 377 | 83 | |
BC161773.1 | - | 3 - 4 | 413 | 83 | ||
BC171056.1 | - | 2 - 3 | 364 | 83 | ||
BC171062.1 | - | 2 - 3 | 364 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |