Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113882.1 | NP_001107354.1 | 6 - 7 | 859 | 62 | |
GenBank mRNA | BC157201.1 | - | 6 - 7 | 859 | 62 | |
BC161773.1 | - | 6 - 7 | 895 | 62 | ||
BC171056.1 | - | 5 - 6 | 846 | 62 | ||
BC171062.1 | - | 5 - 6 | 846 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |