Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001367803.1 | NP_001354732.1 | 4 - 5 | 659 | 91 | |
GenBank mRNA | CU075881.1 | - | 4 - 5 | 659 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |