Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113922.1 | NP_001107394.1 | 8 - 9 | 1122 | 75 | |
GenBank mRNA | BC157332.1 | - | 8 - 9 | 1122 | 75 | |
BC170995.1 | - | 8 - 9 | 1119 | 75 | ||
BC171000.1 | - | 8 - 9 | 1119 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |