Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113951.1 | NP_001107423.1 | 2 - 3 | 204 | 57 | |
XM_012971521.2 | XP_012826975.1 | 2 - 3 | 295 | 57 | ||
GenBank mRNA | BC157553.1 | - | 2 - 3 | 204 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |