Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114065.1 | NP_001107537.1 | 4 - 5 | 431 | 97 | |
GenBank mRNA | BC157789.1 | - | 4 - 5 | 431 | 97 | |
BC170946.1 | - | 4 - 5 | 373 | 97 | ||
BC171296.1 | - | 4 - 5 | 373 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |