Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114206.1 | NP_001107678.1 | 4 - 5 | 370 | 63 | |
GenBank mRNA | BC135180.1 | - | 4 - 5 | 344 | 63 | |
BC153700.1 | - | 4 - 5 | 370 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |