Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fcgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114208.1 | NP_001107680.1 | 4 - 5 | 698 | 76 | |
GenBank mRNA | BC088875.1 | - | 4 - 5 | 670 | 76 | |
BC154883.1 | - | 4 - 5 | 698 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |